COMH4211AC67 Combi-Blocks, Inc.

N-[(2-Methoxynaphthalen-1-yl)methyl]cyclopropanamine

  • MDL number MFCD06761035
  • Cas Number : 880070-23-5
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1703
Availability details for
Quantity
  • Availability
  • Add to Cart