COMH4211AC68 Combi-Blocks, Inc.

5-Chloro-2-(3-fluorophenoxy)nicotinic acid

  • MDL number MFCD31557863
  • Cas Number : 847730-36-3
  • Molecular Weight : 267.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1707

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1707
Availability details for
Quantity
  • Availability
  • Add to Cart