COMH4211AC6B Combi-Blocks, Inc.

2-Isopropoxybenzoyl chloride

  • MDL number MFCD11111058
  • Cas Number : 66849-11-4
  • Molecular Weight : 198.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1715

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1715
Availability details for
Quantity
  • Availability
  • Add to Cart