COMH4211AC6C Combi-Blocks, Inc.

6-[(4-Methoxybenzyl)oxy]-1h-indazol-3-amine

  • MDL number MFCD28975973
  • Cas Number : 1242152-57-3
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1717

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1717
Availability details for
Quantity
  • Availability
  • Add to Cart