COMH4211AC6D Combi-Blocks, Inc.

3-Amino-n-ethyl-5-fluorobenzamide

  • MDL number MFCD24125106
  • Cas Number : 1499211-62-9
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1718
Availability details for
Quantity
  • Availability
  • Add to Cart