COMH4211AC6E Combi-Blocks, Inc.

5-Bromo-2-chloro-n-propyl-nicotinamide

  • MDL number MFCD16159733
  • Cas Number : 1247088-79-4
  • Molecular Weight : 277.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1719

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1719
Availability details for
Quantity
  • Availability
  • Add to Cart