COMH4211AC6F Combi-Blocks, Inc.

(2R)-2-Amino-n-cyclopentyl-3-methylbutanamide

  • MDL number MFCD21234682
  • Cas Number : 1568155-82-7
  • Molecular Weight : 184.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-2-amino-N-cyclopentyl-3-methylbutanamide
Inchi Code 1S/C10H20N2O/c1-7(2)9(11)10(13)12-8-5-3-4-6-8/h7-9H,3-6,11H2,1-2H3,(H,12,13)/t9-/m1/s1
Inchi Key GRJJRKRYXRWSAK-SECBINFHSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1721
Availability details for
Quantity
  • Availability
  • Add to Cart