COMH4211AC70 Combi-Blocks, Inc.

1-((2-(Benzyloxy)ethoxy)methyl)-4-bromo-2-fluorobenzene

  • MDL number MFCD31628755
  • Cas Number : 1402585-62-9
  • Molecular Weight : 339.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1722

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1722
Availability details for
Quantity
  • Availability
  • Add to Cart