COMH4211AC71 Combi-Blocks, Inc.

Ethoxy[(4-fluorophenyl)methyl]amine

  • MDL number MFCD21101274
  • Cas Number : 543730-93-4
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1724

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1724
Availability details for
Quantity
  • Availability
  • Add to Cart