COMH4211AC74 Combi-Blocks, Inc.

[5-(3-Trifluoromethyl-phenyl)-thiophen-2-yl]-methanol

  • MDL number MFCD31619003
  • Cas Number : 1190235-27-8
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1733
Availability details for
Quantity
  • Availability
  • Add to Cart