COMH4211AC75 Combi-Blocks, Inc.

4-Isoxazolemethanol, 3-(2,5-difluorophenyl)-5-methyl-

  • MDL number MFCD22030736
  • Cas Number : 1159602-56-8
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1734
Availability details for
Quantity
  • Availability
  • Add to Cart