COMH4211AC79 Combi-Blocks, Inc.

1-(4-Cyclopropylmethoxy-2-methyl-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31557833
  • Cas Number : 2206609-13-2
  • Molecular Weight : 241.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1745
Availability details for
Quantity
  • Availability
  • Add to Cart