COMH4211AC7A Combi-Blocks, Inc.

1-[2-(4-Fluorophenyl)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD28718224
  • Cas Number : 1430750-42-7
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1747
Availability details for
Quantity
  • Availability
  • Add to Cart