COMH4211AC7D Combi-Blocks, Inc.

1-(4-Isobutoxy-3-nitro-phenyl)-ethanone

  • MDL number MFCD11136890
  • Cas Number : 217485-63-7
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1757
Availability details for
Quantity
  • Availability
  • Add to Cart