COMH4211ACCB Combi-Blocks, Inc.

3-(2-Piperidin-4-yl-ethyl)-benzoic acid methyl ester

  • MDL number MFCD31614019
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1760

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1760
Availability details for
Quantity
  • Availability
  • Add to Cart