COMH4211ACCC Combi-Blocks, Inc.

1-(3,4-Dichlorobenzyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD31583090
  • Cas Number : 1604036-61-4
  • Molecular Weight : 353.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1762

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1762
Availability details for
Quantity
  • Availability
  • Add to Cart