COMH4211ACCF Combi-Blocks, Inc.

(4-Ethoxy-3,5-dimethylphenyl)methanamine

  • MDL number MFCD11192017
  • Cas Number : 1039317-11-7
  • Molecular Weight : 179.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1766
Availability details for
Quantity
  • Availability
  • Add to Cart