COMH4211ACD0 Combi-Blocks, Inc.

4'-Bromo-3'-methyl-biphenyl-3-carboxylic acid methyl ester

  • MDL number MFCD31579461
  • Cas Number : 2270906-41-5
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1767
Availability details for
Quantity
  • Availability
  • Add to Cart