COMH4211ACD1 Combi-Blocks, Inc.

5-Bromo-1,3-difluoro-2-(4-methoxy-benzyloxy)-benzene

  • MDL number MFCD31613883
  • Molecular Weight : 329.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1769

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1769
Availability details for
Quantity
  • Availability
  • Add to Cart