COMH4211ACD5 Combi-Blocks, Inc.

(Trans-4-aminocyclohexyl)-4-morpholinylmethanone

  • MDL number MFCD24641450
  • Cas Number : 412291-29-3
  • Molecular Weight : 212.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1781
Availability details for
Quantity
  • Availability
  • Add to Cart