COMH4211ACD6 Combi-Blocks, Inc.

3-(Propane-2-sulfonyl)-propylamine hydrochloride

  • MDL number MFCD31561553
  • Cas Number : 2270906-40-4
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1792
Availability details for
Quantity
  • Availability
  • Add to Cart