COMH4211ACD9 Combi-Blocks, Inc.

5-Bromo-4-ethylpyridin-2-amine

  • MDL number MFCD22058625
  • Cas Number : 1368352-40-2
  • Molecular Weight : 201.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1802

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1802
Availability details for
Quantity
  • Availability
  • Add to Cart