COMH4211ACDA Combi-Blocks, Inc.

2-(4-Bromo-2-fluoro-phenyl)-n-ethyl-acetamide

  • MDL number MFCD31613955
  • Cas Number : 1788988-87-3
  • Molecular Weight : 260.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-bromo-2-fluorophenyl)-N-ethylacetamide
Inchi Code 1S/C10H11BrFNO/c1-2-13-10(14)5-7-3-4-8(11)6-9(7)12/h3-4,6H,2,5H2,1H3,(H,13,14)
Inchi Key MYAXBPXPZYAWHQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1806
Availability details for
Quantity
  • Availability
  • Add to Cart