COMH4211ACDB Combi-Blocks, Inc.

2-Hydroxy-2'-trifluoromethyl-biphenyl-4-carboxylic acid methyl ester

  • MDL number MFCD31613928
  • Cas Number : 1243559-24-1
  • Molecular Weight : 296.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-hydroxy-2'-(trifluoromethyl)-[1,1'-biphenyl]-4-carboxylate
Inchi Code 1S/C15H11F3O3/c1-21-14(20)9-6-7-11(13(19)8-9)10-4-2-3-5-12(10)15(16,17)18/h2-8,19H,1H3
Inchi Key HMEQKCXDRCDSCC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1807
Availability details for
Quantity
  • Availability
  • Add to Cart