COMH4211ACDC Combi-Blocks, Inc.

Benzyl cis-4-hydroxycyclohexanecarboxylate

  • MDL number MFCD29917336
  • Cas Number : 1369414-98-1
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1813

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1813
Availability details for
Quantity
  • Availability
  • Add to Cart