COMH4211ACE0 Combi-Blocks, Inc.

Cis-benzyl 4-aminocyclohexanecarboxylate hydrochloride

  • MDL number MFCD28383627
  • Cas Number : 67299-44-9
  • Molecular Weight : 269.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1819
Availability details for
Quantity
  • Availability
  • Add to Cart