COMH4211ACE2 Combi-Blocks, Inc.

Trans 3-ethanesulfonylamino-cyclohexanecarboxylic acid

  • MDL number MFCD31618924
  • Cas Number : 2165711-41-9
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1825

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1825
Availability details for
Quantity
  • Availability
  • Add to Cart