COMH4211ACE4 Combi-Blocks, Inc.

3-(4-(Trifluoromethoxy)phenyl)-1,2,4-oxadiazole-5-carbohydrazide

  • MDL number MFCD02679588
  • Cas Number : 883028-82-8
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1833

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1833
Availability details for
Quantity
  • Availability
  • Add to Cart