COMH4211ACE5 Combi-Blocks, Inc.

N-(5-Amino-pyridin-2-yl)-2-phenyl-acetamide

  • MDL number MFCD11182702
  • Cas Number : 1094261-69-4
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1836

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1836
Availability details for
Quantity
  • Availability
  • Add to Cart