COMH4211ACE6 Combi-Blocks, Inc.

(R)-1-(3-(Cyclopropylmethoxy)phenyl)ethanamine hydrochloride

  • MDL number MFCD29085474
  • Cas Number : 1198223-50-5
  • Molecular Weight : 227.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1837

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1837
Availability details for
Quantity
  • Availability
  • Add to Cart