COMH4211ACE7 Combi-Blocks, Inc.

1-(2-Chloro-benzyl)-3,5-dimethyl-1h-pyrazol-4-ol

  • MDL number MFCD21254725
  • Cas Number : 1492674-57-3
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1840

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1840
Availability details for
Quantity
  • Availability
  • Add to Cart