COMH4211ACE9 Combi-Blocks, Inc.

4-Bromo-2-methyl-1-(2-methylpropoxy)benzene

  • MDL number MFCD18361004
  • Cas Number : 1309933-10-5
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1844

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1844
Availability details for
Quantity
  • Availability
  • Add to Cart