COMH4211ACEB Combi-Blocks, Inc.

2-tert-Butoxycarbonylamino-3-(4-trifluoromethyl-phenyl)-acrylic acid methyl ester

  • MDL number MFCD28383623
  • Cas Number : 213741-30-1
  • Molecular Weight : 345.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1847

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1847
Availability details for
Quantity
  • Availability
  • Add to Cart