COMH4211ACEC Combi-Blocks, Inc.

4-(2-Chlorophenyl)-1h-pyrazole

  • MDL number MFCD12756544
  • Cas Number : 91486-39-4
  • Molecular Weight : 178.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1850

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1850
Availability details for
Quantity
  • Availability
  • Add to Cart