COMH4211ACEE Combi-Blocks, Inc.

2,5-Dioxopyrrolidin-1-yl 4-oxopentanoate

  • MDL number MFCD29071399
  • Cas Number : 334616-59-0
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1854
Availability details for
Quantity
  • Availability
  • Add to Cart