COMH4211ACEF Combi-Blocks, Inc.

4-Bromo-2'-methoxybiphenyl

  • MDL number MFCD07775604
  • Cas Number : 169691-87-6
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1855

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1855
Availability details for
Quantity
  • Availability
  • Add to Cart