COMH4211ACF1 Combi-Blocks, Inc.

tert-Butyl 2-(4-formyl-2-methoxyphenoxy)acetate

  • MDL number MFCD09192271
  • Cas Number : 179002-91-6
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1858

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1858
Availability details for
Quantity
  • Availability
  • Add to Cart