COMH4211ACF3 Combi-Blocks, Inc.

Methyl 4-acetyl-3-chlorobenzoate

  • MDL number MFCD27920811
  • Cas Number : 57542-71-9
  • Molecular Weight : 212.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1861

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1861
Availability details for
Quantity
  • Availability
  • Add to Cart