COMH4211ACF4 Combi-Blocks, Inc.

2-Fluoro-4-piperazin-1-yl-benzamide dihydrochloride

  • MDL number MFCD31583024
  • Cas Number : 2270906-96-0
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1866

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1866
Availability details for
Quantity
  • Availability
  • Add to Cart