COMH4211ACF5 Combi-Blocks, Inc.

2-[4-(2,2,2-Trifluoro-ethoxy)-benzyl]-butyric acid

  • MDL number MFCD31583265
  • Cas Number : 2270909-11-8
  • Molecular Weight : 276.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1869

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1869
Availability details for
Quantity
  • Availability
  • Add to Cart