COMH4211ACF6 Combi-Blocks, Inc.

2-(4-Isopropoxy-pyrazol-1-yl)-phenylamine

  • MDL number MFCD31579541
  • Cas Number : 1876791-05-7
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1870

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1870
Availability details for
Quantity
  • Availability
  • Add to Cart