COMH4211ACF7 Combi-Blocks, Inc.

5-Bromo-2-(isopropylsulfonyl)pyridine

  • MDL number MFCD26395221
  • Cas Number : 98626-94-9
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1871

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1871
Availability details for
Quantity
  • Availability
  • Add to Cart