COMH4211ACF8 Combi-Blocks, Inc.

5-(Cyclopropylmethoxy)-2-fluoropyridine

  • MDL number MFCD24190867
  • Cas Number : 213765-91-4
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1873
Availability details for
Quantity
  • Availability
  • Add to Cart