COMH4211ACFA Combi-Blocks, Inc.

(6-Bromo-pyridin-3-yloxy)-acetic acid ethyl ester

  • MDL number MFCD30830165
  • Cas Number : 1835219-93-6
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1879
Availability details for
Quantity
  • Availability
  • Add to Cart