COMH4211ACFB Combi-Blocks, Inc.

N-(3-Chloro-4-fluoro-benzyl)-o-methyl-hydroxylamine

  • MDL number MFCD24288707
  • Cas Number : 543730-53-6
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1881

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1881
Availability details for
Quantity
  • Availability
  • Add to Cart