COMH4211ACFC Combi-Blocks, Inc.

1-(2,5-Dichloro-benzyl)-1h-pyrazol-4-ol

  • MDL number MFCD31561533
  • Cas Number : 1603010-00-9
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1883

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1883
Availability details for
Quantity
  • Availability
  • Add to Cart