COMH4211ACFD Combi-Blocks, Inc.

4-Bromo-3-(2,2,2-trifluoro-ethoxy)-benzoic acid methyl ester

  • MDL number MFCD29917437
  • Cas Number : 1255707-64-2
  • Molecular Weight : 313.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1886

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1886
Availability details for
Quantity
  • Availability
  • Add to Cart