COMH4211ACFF Combi-Blocks, Inc.

1-[(2,5-Difluorophenyl)methyl]-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD28718179
  • Cas Number : 1450642-71-3
  • Molecular Weight : 348.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1888
Availability details for
Quantity
  • Availability
  • Add to Cart