COMH4211AD01 Combi-Blocks, Inc.

3-Chloro-2-(2-methylpropoxy)aniline

  • MDL number MFCD11132858
  • Cas Number : 130566-35-7
  • Molecular Weight : 199.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1894
Availability details for
Quantity
  • Availability
  • Add to Cart