COMH4211AD02 Combi-Blocks, Inc.

4-(3-(Ethoxymethyl)piperidin-1-yl)aniline

  • MDL number MFCD29995674
  • Cas Number : 2097955-32-1
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1895

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1895
Availability details for
Quantity
  • Availability
  • Add to Cart